missing translation for 'onlineSavingsMsg'
Learn More

2-Bromophenethyl alcohol, 99%

Product Code. 10202472 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10202472 1 g Glass bottle 1g
10028871 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10202472 Supplier Thermo Scientific Acros Supplier No. 350550010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1074-16-4 | C8H9BrO | 201.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1074-16-4
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.06
MDL Number MFCD00093566
InChI Key ADLOWZRDUHSVRU-UHFFFAOYSA-N
Synonym 2-2-bromophenyl ethanol, 2-bromophenethyl alcohol, 2-bromophenethylalcohol, 2-2-bromophenyl ethan-1-ol, benzeneethanol, 2-bromo, o-bromophenethyl alcohol, 2-2-bromophenyl ethyl alcohol, 2-bromobenzeneethanol, 2-bromophenylethanol, pubchem16178
PubChem CID 2734089
IUPAC Name 2-(2-bromophenyl)ethanol
SMILES C1=CC=C(C(=C1)CCO)Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Density 1.4830g/mL
Boiling Point 97.0°C (0.7 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4CH2CH2OH
Refractive Index 1.5755 to 1.5775
Quantity 1 g
Specific Gravity 1.483
Formula Weight 201.06
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Bromophenethyl alcohol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromophenethylalcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.