missing translation for 'onlineSavingsMsg'
Learn More

2-Bromophenol, 98%

Product Code. p-7021681
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11498066

Brand: Thermo Scientific Alfa Aesar A14112.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95-56-7 | C6H5BrO | 173.009 g/mol

2-Bromophenol used as a disinfection byproduct found in chlorinated pool water. Used in the preparation of anti-benzofurobenzofuran diimides. It was also used to study the photodegradation of 2-bromophenol using UV-Vis spectroscopy and HPLC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromophenol used as a disinfection byproduct found in chlorinated pool water. Used in the preparation of anti-benzofurobenzofuran diimides. It was also used to study the photodegradation of 2-bromophenol using UV-Vis spectroscopy and HPLC.

Solubility
Soluble in chloroform and ether. Slightly soluble in water.

Notes
Light sensitive. Store in dark. Store in cool, dry place in tightly closed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-56-7
Molecular Formula C6H5BrO
Molecular Weight (g/mol) 173.009
MDL Number MFCD00002146
InChI Key VADKRMSMGWJZCF-UHFFFAOYSA-N
Synonym o-bromophenol, phenol, 2-bromo, bromophenol, 2-bromfenol, 2-bromo phenol, 2-bromo-phenol, phenol, o-bromo, o-bromo-phenol, phenol, bromo, 2-bromfenol czech
PubChem CID 7244
IUPAC Name 2-bromophenol
SMILES C1=CC=C(C(=C1)O)Br

Specifications

Melting Point 5°C
Density 1.636
Boiling Point 194°C to 195°C
Flash Point 92°C (192°F)
Odor Phenol-like
Refractive Index 1.589
Quantity 25 g
UN Number UN3082
Beilstein 1905115
Sensitivity Light sensitive
Merck Index 14,1428
Solubility Information Soluble in chloroform and ether. Slightly soluble in water.
Formula Weight 173.01
Percent Purity 98%
Chemical Name or Material 2-Bromophenol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.