missing translation for 'onlineSavingsMsg'
Learn More

2-Bromophenylboronic acid, 97%

Product Code. 10680915 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10680915 5 g Glass bottle 5g
10778663 25 g Glass bottle 25g
11985851 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10680915 Supplier Thermo Scientific Acros Supplier No. 379990050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 244205-40-1 | C6H6BBrO2 | 200.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 244205-40-1
Molecular Formula C6H6BBrO2
Molecular Weight (g/mol) 200.83
MDL Number MFCD01114672
InChI Key PLVCYMZAEQRYHJ-UHFFFAOYSA-N
Synonym 2-bromophenyl boronic acid, o-bromophenylboronic acid, 2-bromobenzeneboronic acid, 2-bromophenylboronicacid, boronic acid, 2-bromophenyl, bromobenzeneboronic acid, pubchem13095, 2-bromo phenyl boronic acid, 2-bromo-phenyl boronic acid
PubChem CID 2773294
IUPAC Name (2-bromophenyl)boronic acid
SMILES OB(O)C1=CC=CC=C1Br

Specifications

Melting Point 109.0°C to 114.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula BrC6H4B(OH)2
Quantity 5 g
Formula Weight 200.83
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Bromophenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromophenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.