missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoterephthalic acid, 97%

Product Code. 11486997
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11486997 100 g 100g
11466997 5 g 5g
11476997 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11486997 Supplier Thermo Scientific Alfa Aesar Supplier No. A14393.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 586-35-6 | C8H5BrO4 | 245.028 g/mol

2-Bromoterephthalic acid is used as a biochemical for proteomics research. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoterephthalic acid is used as a biochemical for proteomics research. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 586-35-6
Molecular Formula C8H5BrO4
Molecular Weight (g/mol) 245.028
MDL Number MFCD00002403
InChI Key QPBGNSFASPVGTP-UHFFFAOYSA-N
Synonym bromoterephthalic acid, 2-bromobenzene-1,4-dicarboxylic acid, 2-bromoterephthalicacid, 2-bromo-terephthalic acid, 2-bromo-1,4-benzenedicarboxylic acid, 1,4-benzenedicarboxylic acid, 2-bromo, labotest-bb lt00454996, pubchem12798, 2-bromo terephthalic acid, acmc-1aoy9
PubChem CID 68513
IUPAC Name 2-bromoterephthalic acid
SMILES C1=CC(=C(C=C1C(=O)O)Br)C(=O)O

Specifications

Melting Point 297°C to 300°C
Quantity 100 g
UN Number UN2811
Beilstein 2616163
Solubility Information Soluble in water.
Formula Weight 245.03
Percent Purity 97%
Chemical Name or Material 2-Bromoterephthalic acid

RUO – Research Use Only

Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.