missing translation for 'onlineSavingsMsg'
Learn More

2-Bromothioanisole, 98%

Product Code. p-7021339
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11460204

Brand: Thermo Scientific Alfa Aesar A13632.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19614-16-5 | C7H7BrS | 203.097 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19614-16-5
Molecular Formula C7H7BrS
Molecular Weight (g/mol) 203.097
MDL Number MFCD00000066
InChI Key ALAQDUSTXPEHMH-UHFFFAOYSA-N
Synonym 2-bromothioanisole, 2-bromo thioanisole, 1-bromo-2-methylthio benzene, o-bromothioanisole, 1-bromo-2-methylsulfanyl benzene, benzene, 1-bromo-2-methylthio, 2-bromophenyl methyl sulphide, o-bromo methylthio benzene, 2-bromophenyl methyl sulfide, 1-bromo-2-methylsulfanyl-benzene
PubChem CID 88166
IUPAC Name 1-bromo-2-methylsulfanylbenzene
SMILES CSC1=CC=CC=C1Br

Specifications

Melting Point -24°C
Density 1.524
Boiling Point 140°C to 141°C (20 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.633
Quantity 1 g
Beilstein 2243716
Formula Weight 203.11
Percent Purity 98%
Chemical Name or Material 2-Bromothioanisole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.