missing translation for 'onlineSavingsMsg'
Learn More

2-Bromothioanisole, 98%, ACROS Organics™

Product Code. 10698201 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10698201 1 g Glass bottle 1g
10529643 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10698201 Supplier Thermo Scientific Acros Supplier No. 303610010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 19614-16-5
Molecular Formula C7H7BrS
Molecular Weight (g/mol) 203.097
MDL Number MFCD00000066
InChI Key ALAQDUSTXPEHMH-UHFFFAOYSA-N
Synonym 2-bromothioanisole, 2-bromo thioanisole, 1-bromo-2-methylthio benzene, o-bromothioanisole, 1-bromo-2-methylsulfanyl benzene, benzene, 1-bromo-2-methylthio, 2-bromophenyl methyl sulphide, o-bromo methylthio benzene, 2-bromophenyl methyl sulfide, 1-bromo-2-methylsulfanyl-benzene
PubChem CID 88166
IUPAC Name 1-bromo-2-methylsulfanylbenzene
SMILES CSC1=CC=CC=C1Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula BrC6H4SCH3
Refractive Index 1.6320 to 1.6340 (20°C, 589nm)
Quantity 1 g
Formula Weight 203.11
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 2-Bromothioanisole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.