missing translation for 'onlineSavingsMsg'
Learn More

2-Bromothioanisole, 98%

Product Code. 11480204
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11480204 25 g 25g
11460204 1 g 1g
11470204 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480204 Supplier Thermo Scientific Alfa Aesar Supplier No. A13632.14
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 19614-16-5 | C7H7BrS | 203.097 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19614-16-5
Molecular Formula C7H7BrS
Molecular Weight (g/mol) 203.097
MDL Number MFCD00000066
InChI Key ALAQDUSTXPEHMH-UHFFFAOYSA-N
Synonym 2-bromothioanisole, 2-bromo thioanisole, 1-bromo-2-methylthio benzene, o-bromothioanisole, 1-bromo-2-methylsulfanyl benzene, benzene, 1-bromo-2-methylthio, 2-bromophenyl methyl sulphide, o-bromo methylthio benzene, 2-bromophenyl methyl sulfide, 1-bromo-2-methylsulfanyl-benzene
PubChem CID 88166
IUPAC Name 1-bromo-2-methylsulfanylbenzene
SMILES CSC1=CC=CC=C1Br

Specifications

Melting Point -24°C
Density 1.524
Boiling Point 140°C to 141°C (20 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.633
Quantity 25 g
Beilstein 2243716
Formula Weight 203.11
Percent Purity 98%
Chemical Name or Material 2-Bromothioanisole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.