missing translation for 'onlineSavingsMsg'
Learn More

2-Butanone oxime, 99%

Product Code. 10391953 Shop All Thermo Scientific Products
Click to view available options
Quantity:
25 g
100 g
1 kg
Unit Size:
100g
1kg
25g
This item is not returnable. View return policy

Product Code. 10391953

Brand: Thermo Scientific 403330250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 96-29-7 | C4H9NO | 87.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-29-7
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00013935
InChI Key WHIVNJATOVLWBW-SNAWJCMRSA-N
Synonym mek-oxime, butanone oxime, methyl ethyl ketone oxime, 2-butanone oxime, ethyl methyl ketoxime, usaf ek-906, ethyl-methylketonoxim, unii-51yge935u9, 2-butanone, oxime, methyl ethyl ketoxime
PubChem CID 5324276
IUPAC Name (NZ)-N-butan-2-ylidenehydroxylamine
SMILES CC\C(C)=N\O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -30.0°C
Color Colorless to Yellow
Density 0.9230g/mL
Boiling Point 72.0°C (25.0 mmHg)
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CNOHC2H5
Refractive Index 1.4410 to 1.4440
Quantity 25 g
Beilstein 01, IV, 3250
Specific Gravity 0.923
Solubility Information Solubility in water: 114g/L water (20°C).
Viscosity 15 mPa.s (20°C)
Formula Weight 87.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Butanone oxime, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Butanone oxime
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity after single exposure Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H336-May cause drowsiness or dizziness.
  • H350-May cause cancer.
  • H370-Causes damage to organs.
  • H373-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.