missing translation for 'onlineSavingsMsg'
Learn More

(2-Butenyl)triphenylphosphonium chloride, 97%

Product Code. 11340074
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11340074 25 g 25g
11330074 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11340074 Supplier Thermo Scientific Alfa Aesar Supplier No. L00502.14
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 13138-25-5 | C22H22ClP | 352.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13138-25-5
Molecular Formula C22H22ClP
Molecular Weight (g/mol) 352.84
MDL Number MFCD00051868
InChI Key YYTDJYJBYMQMDI-SQQVDAMQSA-M
Synonym 2-butenyl triphenylphosphonium chloride, but-2-en-1-yltriphenylphosphonium chloride, 2e-but-2-en-1-yltriphenylphosphanium chloride
PubChem CID 11824356
IUPAC Name [(E)-but-2-enyl]-triphenylphosphanium;chloride
SMILES [Cl-].C\C=C\C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∽225°C to 230°C (decomposition)
Quantity 25 g
Beilstein 4170527
Sensitivity Hygroscopic
Formula Weight 352.84
Percent Purity 97%
Chemical Name or Material (2-Butenyl)triphenylphosphonium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.