missing translation for 'onlineSavingsMsg'
Learn More

2-Butyne-1,4-diol, 98+%

Product Code. 11471908
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
This item is not returnable. View return policy

Product Code. 11471908

Brand: Thermo Scientific Alfa Aesar A14539.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-65-6 | C4H6O2 | 86.09 g/mol

2-Butyne-1,4-diol is used as a precursor to prepare 1,4-butanediol, 2-butene-1,4-diol and mucochloric acid. It is also used in textile additives, corrosion inhibitors, plasticizers, synthetic resins and polyurethanes. It is an important raw material of vitamin B6. Further, it is used for brightening, preserving and inhibiting nickel plating. In addition, it is used in biological studies for nematocidal activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Butyne-1,4-diol is used as a precursor to prepare 1,4-butanediol, 2-butene-1,4-diol and mucochloric acid. It is also used in textile additives, corrosion inhibitors, plasticizers, synthetic resins and polyurethanes. It is an important raw material of vitamin B6. Further, it is used for brightening, preserving and inhibiting nickel plating. In addition, it is used in biological studies for nematocidal activity.

Solubility
Soluble in water, ethanol, acetone, methanol. Slightly soluble in ethyl ether and chloroform. Insoluble in benzene.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, acid chlorides, acid anhydrides, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-65-6
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00002915
InChI Key DLDJFQGPPSQZKI-UHFFFAOYSA-N
Synonym 2-butyne-1,4-diol, butynediol, 1,4-dihydroxy-2-butyne, 1,4-butynediol, bis hydroxymethyl acetylene, 2-butynediol, 2-butin-1,4-diol, agrisynth b3d, unii-axh202fpqm, 1,4-butynediol van
PubChem CID 8066
ChEBI CHEBI:16413
IUPAC Name but-2-yne-1,4-diol
SMILES C(C#CCO)O

Specifications

Melting Point 54°C to 58°C
Boiling Point 194°C (100 mmHg)
Flash Point 152°C (305°F)
Linear Formula HOCH2C≡CCH2OH
Quantity 250 g
UN Number UN2716
Beilstein 1071237
Solubility Information Soluble in water,ethanol,acetone,methanol. Slightly soluble in ethyl ether and chloroform. Insoluble in benzene.
Formula Weight 86.09
Percent Purity ≥98%
Chemical Name or Material 2-Butyne-1,4-diol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.