missing translation for 'onlineSavingsMsg'
Learn More

2-Carboxycinnamic acid, 98%, ACROS Organics™

Product Code. 10574911 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10574911 25 g 25g
10285571 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10574911 Supplier Thermo Scientific Acros Supplier No. 335020250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 612-40-8
Molecular Formula C10H8O4
Molecular Weight (g/mol) 192.17
MDL Number MFCD00004380
InChI Key SCWPNMHQRGNQHH-AATRIKPKSA-N
Synonym 2-carboxycinnamic acid, 2-2-carboxyvinyl benzoic acid, 2-2-carboxyethenyl benzoic acid, e-2-2-carboxyvinyl benzoic acid, 2-e-3-hydroxy-3-oxoprop-1-enyl benzoic acid, 2-1e-2-carboxyvinyl benzoic acid, 2-e-2-carboxyethenyl benzoic acid, 2-1e-2-carboxyeth-1-en-1-yl benzoic acid, 2-e-2-carboxyvinyl benzoicacid, 2-carboxy cinnamic acid
PubChem CID 904938
IUPAC Name 2-[(1E)-2-carboxyeth-1-en-1-yl]benzoic acid
SMILES OC(=O)\C=C\C1=CC=CC=C1C(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 197°C to 202°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Quantity 25 g
Formula Weight 192.17
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 2-Carboxycinnamic acid, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.