missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1-tritylimidazole, 97%, Thermo Scientific Chemicals

Product Code. 15428347
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15428347 250 mg 250mg
15418347 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15428347 Supplier Thermo Scientific Alfa Aesar Supplier No. H51861.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

2-Chloro-1-tritylimidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-1-tritylimidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67478-48-2
Molecular Formula C22H17ClN2
Molecular Weight (g/mol) 344.842
MDL Number MFCD02179527
InChI Key JTMBWVKMAGHXKI-UHFFFAOYSA-N
Synonym 2-chloro-1-trityl-1h-imidazole, 2-chloro-1-triphenylmethyl imidazole, pubchem8989, acmc-209nza, 2-chloro-1-trityl-imidazole, 1h-imidazole,2-chloro-1-triphenylmethyl
PubChem CID 2773337
IUPAC Name 2-chloro-1-tritylimidazole
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)N4C=CN=C4Cl

Specifications

Melting Point 200°C to 202°C
Quantity 250 mg
Solubility Information Insoluble in water.
Formula Weight 344.84
Percent Purity 97%
Chemical Name or Material 2-Chloro-1-tritylimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.