missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-fluorobenzeneboronic acid, 95%, Thermo Scientific™

Product Code. 15499347
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15499347 250 mg 250mg
15489347 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15499347 Supplier Thermo Scientific Alfa Aesar Supplier No. H52591.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Chloro-3-fluorobenzeneboronic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 871329-52-1
Molecular Formula C6H5BClFO2
Molecular Weight (g/mol) 174.36
MDL Number MFCD07363741
InChI Key TYOGIUGNZOBBHE-UHFFFAOYSA-N
Synonym 2-chloro-3-fluorophenyl boronic acid, 1-borono-2-chloro-3-fluorobenzene, 2-chloro-3-fluorobenzeneboronic acid, pubchem19756, acmc-209qfx, 2-chloro-3-fluoro-phenylboronic acid, 2-chloro-3-fluoro-benzeneboronic acid, 2-chloro-3-fluoro-phenyl boronic acid, 2-chloro-3-fluorophenyl ;boronic acid
PubChem CID 22309459
IUPAC Name (2-chloro-3-fluorophenyl)boronic acid
SMILES OB(O)C1=C(Cl)C(F)=CC=C1

Specifications

Melting Point 80°C to 84°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 174.37
Percent Purity 95%
Chemical Name or Material 2-Chloro-3-fluorobenzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.