missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-fluoropyridine-6-carboxylic acid, 98%

Product Code. 15452039
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15452039 1 g 1g
15459116 250 mg 250mg
15462039 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15452039 Supplier Thermo Scientific Alfa Aesar Supplier No. H64734.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 860296-24-8 | C6H3ClFNO2 | 175.543 g/mol

It is an important intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 860296-24-8
Molecular Formula C6H3ClFNO2
Molecular Weight (g/mol) 175.543
MDL Number MFCD13185819
InChI Key JHFKXHUKNYFWJM-UHFFFAOYSA-N
Synonym 6-chloro-5-fluoropicolinic acid, 2-chloro-3-fluoropyridine-6-carboxylicacid, 2-chloro-3-fluoropyridine-6-carboxylic acid, 6-chloro-5-fluoro-pyridine-2-carboxylic acid, 2-chloro-3-fluoro-6-carboxypyridine, 2-chloro-3-fluoro-6-pyridinecarboxylic acid
PubChem CID 11412672
IUPAC Name 6-chloro-5-fluoropyridine-2-carboxylic acid
SMILES C1=CC(=NC(=C1F)Cl)C(=O)O

Specifications

Color White
Quantity 1 g
Beilstein 10063182
Solubility Information Slightly soluble in water.
Formula Weight 175.54
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Chloro-3-fluoropyridine-6-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.