missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-methoxybenzoic acid, 97%

Product Code. 15595996 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15595996 1 g 1g
15506006 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15595996 Supplier Thermo Scientific Acros Supplier No. 459900010
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 33234-36-5 | C8H7ClO3 | 186.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33234-36-5
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.59
MDL Number MFCD07784399
InChI Key VIUDWLLKFANPLX-UHFFFAOYSA-N
Synonym benzoic acid, 2-chloro-3-methoxy, acmc-20a6cl, 2-chloro-m-anisic acid, 2-chloro-3-methoxybenzoicacid, 2-chloro-3-methoxybenzoic acid
PubChem CID 16218063
IUPAC Name 2-chloro-3-methoxybenzoic acid
SMILES COC1=CC=CC(C(O)=O)=C1Cl

Specifications

Melting Point 161.0°C to 165.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 186.6
Percent Purity 97%
Chemical Name or Material 2-Chloro-3-methoxybenzoic acid
compliance-icons
Product Identifier
  • 2-Chloro-3-methoxybenzoic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.