missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(4-morpholinylcarbonyl)benzeneboronic acid, 97%, Thermo Scientific™

Product Code. 15432046
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15432046 1 g 1g
15442046 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15432046 Supplier Thermo Scientific Alfa Aesar Supplier No. H55340.03
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2000187-73-3
Molecular Formula C11H13BClNO4
Molecular Weight (g/mol) 269.488
MDL Number MFCD22125139
InChI Key OQKJPQSIYRSBBW-UHFFFAOYSA-N
Synonym 2-chloro-4-4-morpholinylcarbonyl benzeneboronic acid, 2-chloro-4-4-morpholinylcarbonyl phenylboronic acid, 2-chloro-4-morpholine-4-carbonyl phenylboronic acid
PubChem CID 73996122
IUPAC Name [2-chloro-4-(morpholine-4-carbonyl)phenyl]boronic acid
SMILES B(C1=C(C=C(C=C1)C(=O)N2CCOCC2)Cl)(O)O

Specifications

Quantity 1 g
Formula Weight 269.49
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-(4-morpholinylcarbonyl)benzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.