missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4,5-dimethylphenol, 98%, Thermo Scientific Chemicals

Product Code. 11331357
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11331357 5 g 5g
11321357 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11331357 Supplier Thermo Scientific Alfa Aesar Supplier No. L01627.06
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1124-04-5
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.609
MDL Number MFCD00002173
InChI Key PSOJLBXHRBFLLQ-UHFFFAOYSA-N
Synonym banol phenol, phenol, 2-chloro-4,5-dimethyl, 2-chloro-4,5-xylenol, 6-chloro-3,4-xylenol, 3,4-dimethyl-6-chlorophenol, 3,4-xylenol, 6-chloro, acmc-20anzv, 3, 6-chloro, dsstox_cid_21404, dsstox_rid_79715
PubChem CID 70754
IUPAC Name 2-chloro-4,5-dimethylphenol
SMILES CC1=CC(=C(C=C1C)Cl)O

Specifications

Melting Point 70°C to 72°C
Quantity 5 g
Beilstein 2042057
Formula Weight 156.61
Percent Purity 98%
Chemical Name or Material 2-Chloro-4,5-dimethylphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.