missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4,6-dimethoxy-1,3,5-triazine, 98%

Product Code. 11415947
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 11415947

Brand: Thermo Scientific Alfa Aesar B24474.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 3140-73-6 | C5H6ClN3O2 | 175.57 g/mol

2-Chloro-4,6-dimethoxy-1,3,5-triazine is used as a peptide coupling agent for the purification of peptides. It is used in the synthesis of 4-(4,6-dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride through coupling with N-methylmorpholine in tetrahydrofuran. Further, it plays an important role in the production of bis(4,6-dimethoxy-1,3,5-triazin-2-yl) ether, through reaction with 2-hydroxy-4,6-dimethoxy-1,3,5-triazine. In addition to this, it is involved in the preparation of 2-(4,6-dimethoxy-1,3,5-triazinyl)trialkylammonium salts through reaction with various tertiary amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-4,6-dimethoxy-1,3,5-triazine is used as a peptide coupling agent for the purification of peptides. It is used in the synthesis of 4-(4,6-dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride through coupling with N-methylmorpholine in tetrahydrofuran. Further, it plays an important role in the production of bis(4,6-dimethoxy-1,3,5-triazin-2-yl) ether, through reaction with 2-hydroxy-4,6-dimethoxy-1,3,5-triazine. In addition to this, it is involved in the preparation of 2-(4,6-dimethoxy-1,3,5-triazinyl)trialkylammonium salts through reaction with various tertiary amines.

Solubility
Soluble in water, chloroform and methanol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3140-73-6
Molecular Formula C5H6ClN3O2
Molecular Weight (g/mol) 175.57
MDL Number MFCD00075607
InChI Key GPIQOFWTZXXOOV-UHFFFAOYSA-N
Synonym cdmt, 1,3,5-triazine, 2-chloro-4,6-dimethoxy, 2-chloro-4,6-dimethoxy-s-triazine, 2-chloro-4,6-dimethoxy-1,3,5 triazine, pubchem20734, acmc-1cnh7, ksc567a8j, 2-chloro-4,6-dimethoxy s-triazine, 1,5-triazine, 2-chloro-4,6-dimethoxy
PubChem CID 18450
IUPAC Name 2-chloro-4,6-dimethoxy-1,3,5-triazine
SMILES COC1=NC(OC)=NC(Cl)=N1

Specifications

Melting Point 75°C to 79°C
Boiling Point 99°C to 100°C (1 mmHg)
Quantity 5 g
Beilstein 148988
Solubility Information Soluble in water,chloroform and methanol.
Formula Weight 175.58
Percent Purity 98%
Chemical Name or Material 2-Chloro-4,6-dimethoxy-1,3,5-triazine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.