missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4,6-diphenyl-1,3,5-triazine, 97%

Product Code. p-7031912
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15495967 1 g 1g
15405977 5 g 5g
15415977 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15495967 Supplier Thermo Scientific Alfa Aesar Supplier No. H33175.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3842-55-5 | C15H10ClN3 | 267.716 g/mol

A rational synthesis of kyaphenine (2:4:6-triphenyl-1:3:5-triazine) has been effected by reaction of 2-chloro-4:6-diphenyl-1:3:5-triazine with phenylmagnesium bromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A rational synthesis of kyaphenine (2:4:6-triphenyl-1:3:5-triazine) has been effected by reaction of 2-chloro-4:6-diphenyl-1:3:5-triazine with phenylmagnesium bromide.

Solubility
Soluble in toluene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3842-55-5
Molecular Formula C15H10ClN3
Molecular Weight (g/mol) 267.716
MDL Number MFCD05738885
InChI Key DDGPPAMADXTGTN-UHFFFAOYSA-N
Synonym 1,3,5-triazine, 2-chloro-4,6-diphenyl, diphenyl chloro-s-triazine, 4,6-diphenyl-2-chloro-s-triazine, acmc-1ctho, 2-chloro4,6-diphenyl-s-triazine, s-triazine, 2-chloro-4,6-diphenyl, 1,5-triazine, 2-chloro-4,6-diphenyl, 2,6-diphenyl-4-chloro-1,3,5-triazine, 1,3,5-riazine, 2-hloro-4,6-iphenyl, 1,3,5-triazine,2-chloro-4,6-diphenyl
PubChem CID 19698
IUPAC Name 2-chloro-4,6-diphenyl-1,3,5-triazine
SMILES C1=CC=C(C=C1)C2=NC(=NC(=N2)Cl)C3=CC=CC=C3

Specifications

Melting Point 138°C to 140°C
Quantity 1 g
Solubility Information Soluble in toluene.
Formula Weight 267.72
Percent Purity 97%
Chemical Name or Material 2-Chloro-4,6-diphenyl-1,3,5-triazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.