missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluoro-5-nitrobenzoic acid, 97%, Thermo Scientific™

Product Code. 15488318
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15488318

Brand: Thermo Scientific Alfa Aesar H61753.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Chloro-4-fluoro-5-nitrobenzoic acid is used as pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114776-15-7
Molecular Formula C7H3ClFNO4
Molecular Weight (g/mol) 219.552
MDL Number MFCD04115715
InChI Key SYZKAFCPWNFONG-UHFFFAOYSA-N
PubChem CID 7210828
SMILES C1=C(C(=CC(=C1[N+](=O)[O-])F)Cl)C(=O)O

Specifications

Melting Point 152°C to 156°C
Color White
Quantity 25 g
Synonym 2-chloro-4-fluoro-5-nitrobenzoic acid, 2-chloro-4-fluoro-5-nitro-benzoic acid, benzoic acid, 2-chloro-4-fluoro-5-nitro, benzoic acid,2-chloro-4-fluoro-5-nitro, acmc-2099ln, 5-carboxy-4-chloro-2-fluoronitrobenzene, 2,2'-bithiophene-5,8'-diylbis trimethylstannane
Solubility Information Soluble in methanol.
Formula Weight 219.55
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Chloro-4-fluoro-5-nitrobenzoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.