missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluorobenzeneboronic acid, 98%

Product Code. 11456500
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11456500

Brand: Thermo Scientific Alfa Aesar H27591.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 313545-72-1 | C6H5BClFO2 | 174.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 313545-72-1
Molecular Formula C6H5BClFO2
Molecular Weight (g/mol) 174.36
MDL Number MFCD02684295
InChI Key XOFNMNLYGPKKOV-UHFFFAOYSA-N
Synonym 2-chloro-4-fluorobenzeneboronic acid, 2-chloro-4-fluorophenyl boronic acid, 2-chloro-4-fluorophenyl-boronic acid, 2-chloro-4-fluoro-phenyl boronic acid, boronic acid, 2-chloro-4-fluorophenyl, pubchem5050, acmc-209hlz, 2-chloro-4-trifluoromethylphenylboronicacid, ksc222e3l, ablock ab-12-5093
PubChem CID 17750229
IUPAC Name (2-chloro-4-fluorophenyl)boronic acid
SMILES OB(O)C1=CC=C(F)C=C1Cl

Specifications

Quantity 1 g
Formula Weight 174.37
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-fluorobenzeneboronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.