missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluorothiophenol, 97%

Product Code. 11485593
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11485593 5 g 5g
11475593 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11485593 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor A13498.06
 Restricted Item
Estimated Shipment: 30-12-2026
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 175277-99-3 | C6H4ClFS | 162.606 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 175277-99-3
Molecular Formula C6H4ClFS
Molecular Weight (g/mol) 162.606
MDL Number MFCD00052936
InChI Key KUAPPJSILOMQPC-UHFFFAOYSA-N
Synonym 2-chloro-4-fluorothiophenol, 2-chloro-4-fluorobenzene-1-thiol, benzenethiol,2-chloro-4-fluoro, 2-chloro-4-fluoro-benzenethiol, benzenethiol, 2-chloro-4-fluoro, pubchem2850, acmc-1caad, 2-chloro-4-fluoro-thiophenol, 4-fluoro-2-chloro-benzenethiol, #
PubChem CID 519446
IUPAC Name 2-chloro-4-fluorobenzenethiol
SMILES C1=CC(=C(C=C1F)Cl)S

Specifications

Boiling Point 86°C to 88°C (15 mmHg)
Refractive Index 1.567
Quantity 5 g
Beilstein 8478226
Sensitivity Air Sensitive
Formula Weight 162.61
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-fluorothiophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.