missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-methylaniline, 98%

Product Code. 11459997
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11459997 25 g 25g
11449997 5 g 5g
11469997 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11459997 Supplier Thermo Scientific Alfa Aesar Supplier No. B22935.14
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 615-65-6 | C7H8ClN | 141.598 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-65-6
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.598
MDL Number MFCD00007666
InChI Key XGYLSRFSXKAYCR-UHFFFAOYSA-N
Synonym 2-chloro-p-toluidine, benzenamine, 2-chloro-4-methyl, 4-amino-3-chlorotoluene, p-toluidine, 2-chloro, 2-chlor-4-toluidin, 4-methyl-2-chloroaniline, unii-k59isj2ko1, 2-chlor-4-toluidin czech, ccris 2887, 3-chloro-4-aminotoluene
PubChem CID 12007
IUPAC Name 2-chloro-4-methylaniline
SMILES CC1=CC(=C(C=C1)N)Cl

Specifications

Melting Point 7°C
Density 1.151
Boiling Point 223°C to 225°C
Flash Point 99°C (210°F)
Refractive Index 1.575
Quantity 25 g
UN Number UN3429
Beilstein 774514
Formula Weight 141.6
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-methylaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.