missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(methylsulfonyl)benzoic acid, 95%, Thermo Scientific™

Product Code. 10683625
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10683625 10 g 10g
10265274 25 g 25g
10406794 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10683625 Supplier Thermo Scientific Maybridge Supplier No. JFD01500EA
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 53250-83-2
Molecular Formula C8H6ClO4S
Molecular Weight (g/mol) 233.64
MDL Number MFCD00216496
InChI Key CTTWSFIIFMWHLQ-UHFFFAOYSA-M
Synonym 2-chloro-4-methylsulfonyl benzoic acid, 2-chloro-4-methylsulphonylbenzoic acid, 2-chloro-4-methanesulfonylbenzoic acid, benzoic acid, 2-chloro-4-methylsulfonyl, 2-chloro-4-methanesulfonyl-benzoic acid, 2-chloro-4-methylsulfonyl benzoic acid dry basis, maybridge1_004133, cmba, pubchem20281, acmc-1aupa
PubChem CID 735863
IUPAC Name 2-chloro-4-methylsulfonylbenzoic acid
SMILES CS(=O)(=O)C1=CC=C(C([O-])=O)C(Cl)=C1

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 234.66
Percent Purity 95%
Chemical Name or Material 2-Chloro-4-(methylsulfonyl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.