missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-nitrophenol, 96%, Thermo Scientific™

Product Code. 10264610 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
Packaging:
Glass bottle
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10264610 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10264610 Supplier Thermo Scientific Acros Supplier No. 109720500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 619-08-9
CAS Min % 95.0
CAS Max % 100.0
Melting Point 106.0°C to 112.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Molecular Formula C6H4ClNO3
Linear Formula ClC6H3(NO2)OH
MDL Number MFCD00043910
Quantity 50 g
Beilstein 06, 240
Synonym 2-chloro-4-nitrophenol, nitrofungin, phenol, 2-chloro-4-nitro, 2-chloro-4-nitro phenol, nitrofurgin, phenol, chloro-4-nitro, unii-y2v03m9ul8, 2-chloro-4-nitro-phenol, 2-chloro-4nitrophenol, pubchem18908
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,ether
InChI Key BOFRXDMCQRTGII-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1[N+](=O)[O-])Cl)O
Molecular Weight (g/mol) 173.56
PubChem CID 12074
Formula Weight 173.56
Percent Purity 96%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Chloro-4-nitrophenol, 96%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-4-nitrophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.