missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-nitrotoluene, 98%

Product Code. 11475557
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475557 100 g 100g
11465557 25 g 25g
11485557 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11475557 Supplier Thermo Scientific Alfa Aesar Supplier No. B23542.22
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 121-86-8 | C7H6ClNO2 | 171.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-86-8
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007210
InChI Key LLYXJBROWQDVMI-UHFFFAOYSA-N
Synonym 2-chloro-4-nitrotoluene, benzene, 2-chloro-1-methyl-4-nitro, o-chloro-p-nitrotoluol, toluene, 2-chloro-4-nitro, 3-chloro-4-methylnitrobenzene, 2-chlor-4-nitrotoluen, unii-9t26lnr6t8, 2-chloro-4-nitrotoluen czech, 2,4-chloronitrotoluene, 2-chloro-4-nitro-1-methylbenzene
PubChem CID 8491
IUPAC Name 2-chloro-1-methyl-4-nitrobenzene
SMILES CC1=C(C=C(C=C1)[N+](=O)[O-])Cl

Specifications

Melting Point 62°C to 66°C
Boiling Point 260°C
Quantity 100 g
UN Number UN3457
Beilstein 1817924
Formula Weight 171.58
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-nitrotoluene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.