missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(trifluoromethyl)aniline, 98%

Product Code. 11484933
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484933 25 g 25g
11474933 5 g 5g
11389429 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11484933 Supplier Thermo Scientific Alfa Aesar Supplier No. A12242.14
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 39885-50-2 | C7H5ClF3N | 195.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39885-50-2
Molecular Formula C7H5ClF3N
Molecular Weight (g/mol) 195.57
MDL Number MFCD00042563
InChI Key MBBUTABXEITVNY-UHFFFAOYSA-N
Synonym 2-chloro-4-trifluoromethyl aniline, 4-amino-3-chlorobenzotrifluoride, 4-amino-5-chloro-benzotrifluoride, benzenamine, 2-chloro-4-trifluoromethyl, 3-chloro-4-aminobenzotrifluoride, 2-chloro-4-trifluoromethyl-phenylamine, 2-chloro-4-trifluoromethyl benzenamine, 2-chloro-4-trifluoromethyl phenylamine, 4-amino-3-chloro-benzotrifluoride, pubchem2474
PubChem CID 162001
IUPAC Name 2-chloro-4-(trifluoromethyl)aniline
SMILES NC1=CC=C(C=C1Cl)C(F)(F)F

Specifications

Density 1.434
Boiling Point 214°C to 218°C
Flash Point 103°C (217°F)
Refractive Index 1.5015
Quantity 25 g
UN Number UN2810
Beilstein 4179703
Formula Weight 195.57
Percent Purity 98%
Chemical Name or Material 2-Chloro-4- (trifluoromethyl)aniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.