missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(trifluoromethyl)benzenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 11349645
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11349645 1 g 1g
11359645 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11349645 Supplier Thermo Scientific Alfa Aesar Supplier No. L19105.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175205-54-6
Molecular Formula C7H3Cl2F3O2S
Molecular Weight (g/mol) 279.05
MDL Number MFCD00052912
InChI Key NJXDBSSSDPOAFI-UHFFFAOYSA-N
Synonym 2-chloro-4-trifluoromethyl benzene-1-sulfonyl chloride, 2-chloro-4-trifluoromethyl benzenesulfonyl chloride, 2-chloro-4-trifluoromethyl benzenesulphonylchloride, 2-chloro-4-trifluoromethyl benzenesulphonyl chloride, 2-chloro-4-trifluoromethyl benzenesulfonylchloride, 2-chloro-4-trifluoromethyl benzenesulfonyl, 2-chloro-4-trifluoromethyl-benzenesulfonyl chloride, benzenesulfonyl chloride, 2-chloro-4-trifluoromethyl, pubchem5104, acmc-1bwmt
PubChem CID 2736675
IUPAC Name 2-chloro-4-(trifluoromethyl)benzenesulfonyl chloride
SMILES FC(F)(F)C1=CC(Cl)=C(C=C1)S(Cl)(=O)=O

Specifications

Melting Point 35°C to 39°C
Density 1.6
Boiling Point 70°C to 71°C (0.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.575
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 279.07
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-(trifluoromethyl)benzenesulfonyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.