missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-fluoroaniline, 97%

Product Code. 11359909
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11359909

Brand: Thermo Scientific Alfa Aesar B25618.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 452-83-5 | C6H5ClFN | 145.561 g/mol

2-Chloro-5-fluoroaniline is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-5-fluoroaniline is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 452-83-5
Molecular Formula C6H5ClFN
Molecular Weight (g/mol) 145.561
MDL Number MFCD00084852
InChI Key VWUFOZAFKYOZJB-UHFFFAOYSA-N
Synonym 5-fluoro-2-chloroaniline, 2-chloro-5-fluorobenzenamine, 2-chloro-5-fluoro-phenylamine, 2-chloranyl-5-fluoranyl-aniline, pubchem1517, acmc-1bn9k, 2-chloro-5-fluoro-aniline, 2-chloro-5-fluorophenylamine, ksc235o7b, 2-chloro-5-fluorobenzeneamine
PubChem CID 2736509
IUPAC Name 2-chloro-5-fluoroaniline
SMILES C1=CC(=C(C=C1F)N)Cl

Specifications

Melting Point ∼26°C
Boiling Point 211°C
Quantity 5 g
Odor Characteristic
Refractive Index 1.556
UN Number UN2811
Solubility Information Insoluble in water.
Formula Weight 145.56
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-fluoroaniline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.