missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-fluorobenzeneboronic acid, 98%, Thermo Scientific™

Product Code. 15489677
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15489677 1 g 1g
15499677 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15489677 Supplier Thermo Scientific Alfa Aesar Supplier No. H52906.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Chloro-5-fluorobenzeneboronic acid is used as pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 444666-39-1
Molecular Formula C6H5BClFO2
Molecular Weight (g/mol) 174.362
MDL Number MFCD06656272
InChI Key OVTZJDSREVDCTJ-UHFFFAOYSA-N
Synonym 2-chloro-5-fluorobenzeneboronic acid, 2-chloro-5-fluorophenyl boronic acid, 2-chloro-5-fluorophenyl-boronic acid, 2-chloro-5-fluoro-phenyl boronic acid, boronic acid, 2-chloro-5-fluorophenyl, 2-chloro-5-fluorobenzeneboronicacid, acmc-1aqt7, ksc494a1h, 2chloro-5-fluorophenylboronic acid, 2-chloro-5-fluoro-phenylboronic acid
PubChem CID 22648116
IUPAC Name (2-chloro-5-fluorophenyl)boronic acid
SMILES B(C1=C(C=CC(=C1)F)Cl)(O)O

Specifications

Melting Point 78°C to 82°C
Quantity 1 g
Solubility Information Soluble in methanol.
Formula Weight 174.37
Percent Purity 98%
Chemical Name or Material 2-Chloro-5-fluorobenzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.