missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-fluorophenol, 98%

Product Code. 15413827
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15413827 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15413827 Supplier Thermo Scientific Alfa Aesar Supplier No. H28567.06
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 3827-49-4 | C6H4ClFO | 146.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3827-49-4
Boiling Point 183°C to 185°C
Odor Phenol-like
Molecular Formula C6H4ClFO
Refractive Index 1.525
MDL Number MFCD00042524
Quantity 5 g
UN Number UN3265
Synonym 2-chloro-5-fluoro-phenol, pubchem2146, 2-chloro-5-fluorphenol, acmc-209iz2, ksc226k0t, attercop-chm at137231
InChI Key CMQOZIKIOASEIN-UHFFFAOYSA-N
SMILES OC1=CC(F)=CC=C1Cl
IUPAC Name 2-chloro-5-fluorophenol
Molecular Weight (g/mol) 146.55
PubChem CID 2724522
Formula Weight 146.55
Percent Purity 98%
Chemical Name or Material 2-Chloro-5-fluorophenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.