missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-fluoropyrimidine, 97%

Product Code. 15441178
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15441178 1 g 1g
15461178 250 mg 250mg
15451178 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15441178 Supplier Thermo Scientific Alfa Aesar Supplier No. H55055.03
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 62802-42-0 | C4H2ClFN2 | 132.522 g/mol

2-Chloro-5-fluoropyrimidine is used to prepare 2,4-disubstituted-5-fluoropyrimidine , which is a biologically active molecule seen in anticancer agents like 5-fluorouracil.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-5-fluoropyrimidine is used to prepare 2,4-disubstituted-5-fluoropyrimidine , which is a biologically active molecule seen in anticancer agents like 5-fluorouracil.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62802-42-0
Molecular Formula C4H2ClFN2
Molecular Weight (g/mol) 132.522
MDL Number MFCD03788197
InChI Key AGYUQBNABXVWMS-UHFFFAOYSA-N
Synonym 2-chloro-5-fluoro-pyrimidine, 5-fluoro-2-chloropyrimidine, pyrimidine, 2-chloro-5-fluoro, 2-chlor-5-fluorpyrimidin, 2-chloro-5-fluoro pyrimidine, pubchem4684, acmc-1b6zl, ksc352s4h, #, 2-chloro-5-fluoro-1,3-diazine
PubChem CID 583466
IUPAC Name 2-chloro-5-fluoropyrimidine
SMILES C1=C(C=NC(=N1)Cl)F

Specifications

Density 1.073
Flash Point 65°C (149°F)
Refractive Index 1.503
Quantity 1 g
UN Number UN3267
Formula Weight 132.52
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-fluoropyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.