missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-iodobenzoic acid, 97%

Product Code. p-7048052
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11378976

Brand: Thermo Scientific Alfa Aesar L20161.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19094-56-5 | C7H4ClIO2 | 282.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19094-56-5
Molecular Formula C7H4ClIO2
Molecular Weight (g/mol) 282.46
MDL Number MFCD00079731
InChI Key GEBYSTBEDVQOTK-UHFFFAOYSA-N
Synonym 2-chloro-5-iodo-benzoic acid, benzoic acid, 2-chloro-5-iodo, pubchem3649, acmc-209eug, 5-iodo-2-chlorobenzoic acid, ksc174i1h, benzoicacid, 2-chloro-5-iodo, buttpark 100\01-43, 2-chloro-5-iodobenzoic acid
PubChem CID 519638
IUPAC Name 2-chloro-5-iodobenzoic acid
SMILES OC(=O)C1=C(Cl)C=CC(I)=C1

Specifications

Melting Point 156°C to 161°C
Quantity 5 g
Beilstein 2254110
Sensitivity Light sensitive
Formula Weight 282.46
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-iodobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.