missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-methoxyaniline, 98+%

Product Code. 11328986
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328986 100 g 100g
11308986 5 g 5g
11318986 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11328986 Supplier Thermo Scientific Alfa Aesar Supplier No. L20162.22
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 2401-24-3 | C7H8ClNO | 157.597 g/mol

2-Chloro-5-methoxyaniline is used in the preparation of 4-amino-5-chloro-2-methoxyphenyl thiocyanate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-5-methoxyaniline is used in the preparation of 4-amino-5-chloro-2-methoxyphenyl thiocyanate.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2401-24-3
Molecular Formula C7H8ClNO
Molecular Weight (g/mol) 157.597
MDL Number MFCD00047830
InChI Key GBOUQGUQUUPGLO-UHFFFAOYSA-N
Synonym 6-chloro-m-anisidine, 3-amino-4-chloroanisole, benzenamine, 2-chloro-5-methoxy, 2-chloro-5-methoxy-aniline, 6-chloro-m-anisidine hydrochloride, 2-chloro-5-methoxy-phenylamine, 2-chloro-5-methoxybenzenamine hydrochloride, 6-chloro-meta-anisidine hydrochloride, 2-chloro-5-methoxyanilinehydrochloride, timtec-bb sbb008038
PubChem CID 75460
IUPAC Name 2-chloro-5-methoxyaniline
SMILES COC1=CC(=C(C=C1)Cl)N

Specifications

Melting Point 27°C
Boiling Point 135°C to 137°C (12 mmHg)
Odor Characteristic
Refractive Index 1.5865
Quantity 100 g
UN Number UN2233
Beilstein 2082193
Solubility Information Slightly soluble in water.
Formula Weight 157.6
Percent Purity ≥98%
Chemical Name or Material 2-Chloro-5-methoxyaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.