missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-methylaniline, 98%

Product Code. 11386629
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11386629 5 g 5g
11396629 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11386629 Supplier Thermo Scientific Alfa Aesar Supplier No. H26209.06
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 95-81-8 | C7H8ClN | 141.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-81-8
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.60
MDL Number MFCD00007674
InChI Key HPSCXFOQUFPEPE-UHFFFAOYSA-N
Synonym benzenamine, 2-chloro-5-methyl, 6-chloro-m-toluidine, 3-amino-4-chlorotoluene, 2-chlor-5-methyl-aniline, 2-chloro-5-methylbenzenamine, 2-chloro-5-methylphenylamine, ccris 2888, 2-chloro-5-methyl aniline, 6-chlor-meta-toluidin, pubchem18921
PubChem CID 66770
IUPAC Name 2-chloro-5-methylaniline
SMILES CC1=CC=C(Cl)C(N)=C1

Specifications

Melting Point 31°C to 33°C
Boiling Point 228°C to 230°C
Flash Point 107°C (225°F)
Quantity 5 g
UN Number UN2239
Formula Weight 141.6
Percent Purity 98%
Chemical Name or Material 2-Chloro-5-methylaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.