missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-nitronicotinic acid, 95%, Thermo Scientific™

Product Code. 15442839
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15442839 1 g 1g
15452839 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15442839 Supplier Thermo Scientific Alfa Aesar Supplier No. H66927.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42959-38-6
Molecular Formula C6H3ClN2O4
Molecular Weight (g/mol) 202.55
MDL Number MFCD06658402
InChI Key WOFZRBCITDVRON-UHFFFAOYSA-N
Synonym 2-chloro-5-nitronicotinic acid, 2-chloro-5-nitropyridine-3-carboxylic acid, 2-chloro-5-nitro-nicotinic acid, 3-carboxy-2-chloro-5-nitropyridine, 3-carboxy-2-chloro-5-nitro pyridine, 2-chloro-5-nitronicotinicacid, pubchem9631, acmc-209js2, ksc235o0f, chloro-5-nitro nicotinic acid
PubChem CID 12542664
IUPAC Name 2-chloro-5-nitropyridine-3-carboxylic acid
SMILES OC(=O)C1=CC(=CN=C1Cl)[N+]([O-])=O

Specifications

Melting Point 140°C to 143°C
Color White to Yellow
Quantity 1 g
Sensitivity Moisture sensitive
Formula Weight 202.55
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Chloro-5-nitronicotinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.