missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-fluorobenzoyl chloride, 98%, Thermo Scientific™

Product Code. 11460208
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460208 25 g 25g
11450208 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11460208 Supplier Thermo Scientific Alfa Aesar Supplier No. B23762.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79455-63-3
Molecular Formula C7H3Cl2FO
Molecular Weight (g/mol) 192.998
MDL Number MFCD00178746
InChI Key GFNAJZAKJGKJCS-UHFFFAOYSA-N
Synonym 2-chloro-6-fluorobenzoylchloride, 2-chloro-6-fluorobenzene-1-carbonyl chloride, 2-chloro-6-fluoro-benzoyl chloride, 6-chloro-2-fluorobenzoyl chloride, benzoyl chloride, 2-chloro-6-fluoro, acmc-1behg, 2-fluoro-6-chlorobenzoyl chloride, attercop-chm at108371, 2-chloro-6-fluoro benzoyl chloride, 2-chloranyl-6-fluoranyl-benzoyl chloride
PubChem CID 2736547
IUPAC Name 2-chloro-6-fluorobenzoyl chloride
SMILES C1=CC(=C(C(=C1)Cl)C(=O)Cl)F

Specifications

Refractive Index 1.525
Quantity 25 g
UN Number UN3265
Beilstein 4980867
Sensitivity Moisture sensitive
Formula Weight 193.01
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-fluorobenzoyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.