missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-6-fluorobenzyl bromide, 96%, Thermo Scientific™

Product Code. 11954531 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11954531 5 g 5g
11944531 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11954531 Supplier Thermo Scientific Acros Supplier No. 373760050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 68220-26-8
Molecular Formula C7H5BrClF
Molecular Weight (g/mol) 223.47
MDL Number MFCD00040126
InChI Key IGUVNNXFTDCASP-UHFFFAOYSA-N
Synonym 2-chloro-6-fluorobenzyl bromide, 2-bromomethyl-1-chloro-3-fluorobenzene, 2-chloro-6-fluorobenzylbromide, 2-bromomethyl-1-chloro-3-fluoro-benzene, benzene, 2-bromomethyl-1-chloro-3-fluoro, 2-bromomethyl-3-chloro-1-fluorobenzene, alpha-bromo-2-chloro-6-fluorotoluene, pubchem4894, acmc-1bdpq
PubChem CID 2773625
IUPAC Name 2-(bromomethyl)-1-chloro-3-fluorobenzene
SMILES FC1=CC=CC(Cl)=C1CBr

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.6290g/mL
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Linear Formula Cl(F)C6H3CH2Br
Refractive Index 1.569
Quantity 5 g
Specific Gravity 1.629
Formula Weight 223.47
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 2-chloro-6-fluorobenzyl bromide
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-6-fluorobenzyl bromide
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.