missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-fluorobenzyl chloride, 98%, Thermo Scientific™

Product Code. 11449313
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11449313 25 g 25g
11459313 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11449313 Supplier Thermo Scientific Alfa Aesar Supplier No. A11758.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55117-15-2
Molecular Formula C7H5Cl2F
Molecular Weight (g/mol) 179.02
MDL Number MFCD00000896
InChI Key MJGOLNNLNQQIHR-UHFFFAOYSA-N
Synonym 2-chloro-6-fluorobenzyl chloride, 1-chloro-2-chloromethyl-3-fluorobenzene, benzene, 1-chloro-2-chloromethyl-3-fluoro, 2-chloro-6-fluorobenzylchloride, unii-a581kl6zpn, 1-chloro-2-chloromethyl-3-fluoro-benzene, alpha,2-dichloro-6-fluorotoluene, a581kl6zpn, 3-chloro-2-chloromethyl-1-fluorobenzene, pubchem7497
PubChem CID 108675
IUPAC Name 1-chloro-2-(chloromethyl)-3-fluorobenzene
SMILES FC1=CC=CC(Cl)=C1CCl

Specifications

Density 1.401
Boiling Point 208°C to 210°C
Flash Point 93°C (199°F)
Refractive Index 1.537
Quantity 25 g
UN Number UN3265
Beilstein 386840
Formula Weight 179.02
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-fluorobenzyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.