missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methylaniline, Thermo Scientific™

Product Code. 10661745 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10661745 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10661745 Supplier Thermo Scientific Acros Supplier No. 109360100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Spécification

CAS 87-63-8
Melting Point 2.0°C
Density 1.1520g/mL
Boiling Point 215.0°C
Flash Point 98°C
Packaging Glass bottle
Molecular Formula C7H8ClN
Linear Formula ClC6H3(CH3)NH2
Refractive Index 1.5750 to 1.5780
MDL Number MFCD00007679
Quantity 10 g
Beilstein 12,I,388
Specific Gravity 1.152
Synonym 6-chloro-2-methylaniline, 6-chloro-o-toluidine, 2-amino-3-chlorotoluene, benzenamine, 2-chloro-6-methyl, 3-chloro-2-aminotoluene, 6-chloro-2-toluidine, 2-methyl-6-chloroaniline, o-toluidine, 6-chloro, 2-chloro-6-methyl-aniline, 2-chloro-6-methylaninile
Solubility Information Solubility in water: immiscible in water. Other solubilities: miscible with toluene and chlorobenzene
InChI Key WFNLHDJJZSJARK-UHFFFAOYSA-N
SMILES CC1=CC=CC(Cl)=C1N
IUPAC Name 2-chloro-6-methylaniline
Molecular Weight (g/mol) 141.60
PubChem CID 6897
Formula Weight 141.6
Chemical Name or Material 2-Chloro-6-methylaniline
Afficher plus Afficher moins
compliance-icons
Identifiant produit
  • 2-Chloro-6-methylaniline
Mention d'avertissement
  • Warning
Catégorie de danger
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Conseils de prudence
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.