missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methylphenol, 98%

Product Code. p-7043949
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
4.16mL
This item is not returnable. View return policy

Product Code. 11392236

Brand: Thermo Scientific Alfa Aesar L03617.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 87-64-9 | C7H7ClO | 142.582 g/mol

2-Chloro-6-methylphenol is s an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-6-methylphenol is s an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Soluble in water (24 mg/ml at 25°C).

Notes
Storage temperature: 2 - 4°C. Incompatible with Strong oxidizing agents, strong acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-64-9
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.582
MDL Number MFCD00020000
InChI Key YPNZJHFXFVLXSE-UHFFFAOYSA-N
Synonym 6-chloro-o-cresol, phenol, 2-chloro-6-methyl, o-cresol, 6-chloro, 2-methyl-6-chlorophenol, 2-chloro-6-methyl-phenol, 6-chloro cresol, 6-chlor-o-kresol, pubchem3669, 6-chloro-2-methylphenol, 2-chloro-6-methyl phenol
PubChem CID 6898
IUPAC Name 2-chloro-6-methylphenol
SMILES CC1=C(C(=CC=C1)Cl)O

Specifications

Density 1.201
Boiling Point 75°C to 76°C (12 mmHg)
Flash Point 72°C (161°F)
Refractive Index 1.545
Quantity 5 g
UN Number UN2669
Beilstein 774623
Solubility Information Soluble in water (24mg/ml at 25°C).
Formula Weight 142.59
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-methylphenol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-6-methylphenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P370+P378-In case of fire: Use dry sand, dry chemical, or alcohol-resistant foam to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : 6-Chloro-o-cresol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.