missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-nitrotoluene, 98+%, Thermo Scientific™

Product Code. 11430088
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11430088 250 g 250g
11420088 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11430088 Supplier Thermo Scientific Alfa Aesar Supplier No. B23675.30
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-42-1
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007205
InChI Key XCSNRORTQRKCHB-UHFFFAOYSA-N
Synonym 2-chloro-6-nitrotoluene, 6-chloro-2-nitrotoluene, benzene, 1-chloro-2-methyl-3-nitro, toluene, 2-chloro-6-nitro, 3-chloro-2-methylnitrobenzene, caswell no. 202, unii-wez013857x, epa pesticide chemical code 019001, chloro-o-nitrotoluene, pubchem14277
PubChem CID 6740
IUPAC Name 1-chloro-2-methyl-3-nitrobenzene
SMILES CC1=C(Cl)C=CC=C1[N+]([O-])=O

Specifications

Melting Point 34°C to 38°C
Boiling Point 238°C
Flash Point 125°C (257°F)
Quantity 250 g
UN Number UN3457
Beilstein 1239924
Formula Weight 171.58
Percent Purity ≥98%
Chemical Name or Material 2-Chloro-6-nitrotoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.