missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-phenoxybenzylamine, ≥95%, Thermo Scientific™

Product Code. 10702683
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10702683 250 mg Amber Glass Bottle 250mg
10232882 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10702683 Supplier Thermo Scientific Maybridge Supplier No. CD11381CB
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 175136-89-7
Molecular Formula C13H12ClNO
Molecular Weight (g/mol) 233.70
MDL Number MFCD00052915
InChI Key SNTOZVXKDWQFEW-UHFFFAOYSA-N
Synonym 2-chloro-6-phenoxybenzylamine, 2-chloro-6-phenoxyphenyl methanamine, 1-2-chloro-6-phenoxyphenyl methanamine, rarechem al bw 1398, d0x2jg, 2-chloro-6-phenoxy-benzylamine, 2-aminomethyl-3-chlorodiphenylether, 6-chloro-2-phenoxyphenyl methylamine, 2-chloro-6-phenoxybenzylamine, 95+%, benzenemethanamine,2-chloro-6-phenoxy
PubChem CID 2777206
SMILES NCC1=C(OC2=CC=CC=C2)C=CC=C1Cl

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name (2-chloro-6-phenoxyphenyl)methanamine
Formula Weight 233.7
Percent Purity 95%
Chemical Name or Material 2-Chloro-6-phenoxybenzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.