missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroanisole, 98%, ACROS Organics™

Product Code. 10012110 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10012110 50 g Glass bottle 50g
10225700 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10012110 Supplier Thermo Scientific Acros Supplier No. 171420500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 766-51-8
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.582
MDL Number MFCD00000557
InChI Key QGRPVMLBTFGQDQ-UHFFFAOYSA-N
Synonym 2-chloroanisole, o-chloroanisole, benzene, 1-chloro-2-methoxy, anisole, o-chloro, o-chloromethoxybenzene, o-chlorophenyl methyl ether, 2-chlorophenol methyl ether, chloroanisole, 1-chloro-2-methoxy-benzene, labotest-bb ltbb002921
PubChem CID 13011
IUPAC Name 1-chloro-2-methoxybenzene
SMILES COC1=CC=CC=C1Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula ClC6H4OCH3
Refractive Index 1.5435 to 1.5455 (20°C, 589nm)
Quantity 50 g
Beilstein 06, 184
Formula Weight 142.58
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 2-Chloroanisole, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.