missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzaldehyde, 99%

Product Code. 10635592 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
100 mL
250 mL
500 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
100mL
250mL
500mL
5mL
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10635592

Brand: Thermo Scientific Acros 154610010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 89-98-5 | C7H5ClO | 140.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-98-5
Molecular Formula C7H5ClO
Molecular Weight (g/mol) 140.57
MDL Number MFCD00003304
InChI Key FPYUJUBAXZAQNL-UHFFFAOYSA-N
Synonym o-chlorobenzaldehyde, chlorobenzaldehyde, benzaldehyde, 2-chloro, benzaldehyde, o-chloro, 2-chlorbenzaldehyd, 2-clorobenzaldeide, o-chloorbenzaldehyde, usaf m-7, 2-chloorbenzaldehyde, o-chlorobenzenecarboxaldehyde
PubChem CID 6996
IUPAC Name 2-chlorobenzaldehyde
SMILES C1=CC=C(C(=C1)C=O)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 10°C to 11.5°C
Color Colorless to Yellow
Density 1.2500g/mL
Boiling Point 209°C to 215°C
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H4CHO
Refractive Index 1.5650 to 1.567
Quantity 1 L
Beilstein 07, 233
Specific Gravity 1.25
Solubility Information Solubility in water: practically insoluble in water
Formula Weight 140.57
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Chlorobenzaldehyde, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chlorobenzaldehyde
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.