missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorobenzoic acid, 98+%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 138°C to 142°C |
| Density | 1.544 |
| Boiling Point | 284°C to 286°C |
| Flash Point | 173°C (343°F) |
| Quantity | 250 g |
| Beilstein | 907340 |
| Merck Index | 14,2124 |
| Formula Weight | 156.57 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 2-Chlorobenzoic acid |
Safety and Handling
Product Identifier
- 2-Chlorobenzoic acid
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : o-Chlorobenzoic acid
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?