missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzoic Acid, TRC

Product Code. 30005118
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30005118 100 g 100g
17962775 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30005118 Supplier Toronto Research Chemicals Supplier No. TRCC364640100G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-91-2
Molecular Formula C7 H5 Cl O2
Molecular Weight (g/mol) 156.57
Synonym Benzoic acid, o-chloro- (7CI,8CI), 2-Chlorobenzoic acid, 2-CBA, NSC 15042, NSC 32737, o-Chlorobenzoic acid
IUPAC Name 2-chlorobenzoic acid
SMILES OC(=O)c1ccccc1Cl

Specifications

Quantity 100 g
InChI Formula InChI=1S/C7H5ClO2/c8-6-4-2-1-3-5(6)7(9)10/h1-4H,(H,9,10)
Formula Weight 155.9978
Chemical Name or Material 2-Chlorobenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.