missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzoyl chloride, 97%

Product Code. 11482507
Change view
Click to view available options
Quantity:
100 g
250 g
1000 g
Unit Size:
1000g
100g
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11482507 250 g 250g
11389639 100 g 100g
11472507 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11482507 Supplier Thermo Scientific Alfa Aesar Supplier No. A14785.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 609-65-4 | C7H4Cl2O | 175.01 g/mol

2-Chlorobenzoyl chloride, is used in organic synthesis. It is used as a pharmaceutical intermediate for eg as an intermediate of clotrimazolc, preparation of p-chlorobenzoic acid and production of trichloro-acaricide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorobenzoyl chloride, is used in organic synthesis. It is used as a pharmaceutical intermediate for eg as an intermediate of clotrimazolc, preparation of p-chlorobenzoic acid and production of trichloro-acaricide.

Solubility
Soluble in acetone, ether, and alcohol. Solubility in water it decomposes.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Moisture sensitive. Stable under recommended storage conditions. Keep away from strong bases, alcohols, oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-65-4
Molecular Formula C7H4Cl2O
Molecular Weight (g/mol) 175.01
MDL Number MFCD00000660
InChI Key ONIKNECPXCLUHT-UHFFFAOYSA-N
Synonym benzoyl chloride, 2-chloro, o-chlorobenzoyl chloride, chlorobenzoyl chloride, benzoyl chloride, o-chloro, ococ, 2-chloro-benzoyl chloride, unii-jr29a4n74x, o-chlorobenzoylchloride, 2-chlorobenzoylchloride, 2-chlorobenzoic acid chloride
PubChem CID 69110
ChEBI CHEBI:60719
IUPAC Name 2-chlorobenzoyl chloride
SMILES ClC(=O)C1=CC=CC=C1Cl

Specifications

Melting Point -4°C
Density 1.379
Boiling Point 237°C to 239°C
Flash Point 125°C (257°F)
Odor Acrid
Refractive Index 1.5718
Quantity 250 g
UN Number UN3265
Beilstein 386435
Sensitivity Moisture sensitive
Solubility Information Soluble in acetone,ether,and alcohol. Solubility in water it decomposes.
Formula Weight 175.01
Percent Purity 97%
Chemical Name or Material 2-Chlorobenzoyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.