missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzylamine, 97%

Product Code. 10365690 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10365690 25 mL Glass bottle 25mL
10214280 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10365690 Supplier Thermo Scientific Acros Supplier No. 108780250
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 89-97-4 | C7H8ClN | 141.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-97-4
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.6
MDL Number MFCD00008108
InChI Key KDDNKZCVYQDGKE-UHFFFAOYSA-N
Synonym 2-chlorobenzylamine, 2-chlorophenyl methanamine, o-chlorobenzylamine, benzenemethanamine, 2-chloro, o-chlorobenzyl amine, 2-chlorobenzenemethanamine, 2-chloro-benzylamine, benzylamine, o-chloro, 2-chlorophenyl methylamine, unii-l790j4y7a3
PubChem CID 66648
IUPAC Name (2-chlorophenyl)methanamine
SMILES C1=CC=C(C(=C1)CN)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 1.1700g/mL
Boiling Point 103.0°C to 104.0°C (11.0 mmHg)
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H4CH2NH2
Refractive Index 1.5620 to 1.5640
Quantity 25 mL
Beilstein 12, 1073
Specific Gravity 1.17
Solubility Information Solubility in water: immiscible with water
Formula Weight 141.6
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Chlorobenzylamine, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Chlorobenzylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.