missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethyl phenyl sulfone, 98%

Product Code. 11410277
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410277 5 g 5g
11420277 25 g 25g
11430277 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11410277 Supplier Thermo Scientific Alfa Aesar Supplier No. B21690.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 938-09-0 | C8H9ClO2S | 204.668 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 938-09-0
Molecular Formula C8H9ClO2S
Molecular Weight (g/mol) 204.668
MDL Number MFCD00025047
InChI Key NUJGORANFDSMOL-UHFFFAOYSA-N
Synonym 2-chloroethyl phenyl sulfone, 2-chloroethanesulfonyl benzene, 2-chloroethyl sulfonyl benzene, 2-chloroethylphenylsulfone, 2-chloroethylsulfonyl benzene, sulfone, 2-chloroethyl phenyl, 2-chloroethyl sulphonyl benzene, benzene, 2-chloroethyl sulfonyl, 2-chloroethylphenyl sulfone
PubChem CID 13646
IUPAC Name 2-chloroethylsulfonylbenzene
SMILES C1=CC=C(C=C1)S(=O)(=O)CCCl

Specifications

Melting Point 52°C to 55°C
Boiling Point 168°C to 174°C (7 mmHg)
Quantity 5 g
UN Number UN1759
Beilstein 1104341
Formula Weight 204.68
Percent Purity 98%
Chemical Name or Material 2-Chloroethyl phenyl sulfone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.